Learn More
ZNF500 Mouse anti-Human, Clone: OTI1E2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803349
Catalog No. 50-166-6440
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZNF500 is a new candidate transcription factor. It is derived from an annotated genomic sequence (NT_037887) using a gene prediction method.
Specifications
Specifications
ZNF500
Monoclonal
Unconjugated
ZNF500
Lyophilized
ZNF500
Human recombit protein fragment corresponding to amino acids 102-325 of human ZNF500 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1E2
PBS with 8% trehalose and no preservative; pH 7.3
O60304
ZKSCAN18, ZSCAN50
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
26048