Learn More
Aviva Systems Biology Corp ZNF501 Antibody - C-terminal region (ARP50789_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50789_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF501. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF501. Alternate names: MGC21738, ZNF, ZNF52ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-60434