Learn More
ZNF511 Mouse anti-Human, Clone: 3G1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524569
Catalog No. PIMA524569
Add to cart
Description
Description
Peptide Sequence: PLPVERDPAA GAAPFRFVAR PVRFPREHQF FEDGDVQRHL YLQDVIMQVA DVPEKPRVPA FACQVAGCCQ VFDALDDYEH HYHTLHGNVC SFCKRAFPSG HLLDAHILEW HDSLFQILSE RQDMYQCLVE GCTEKFKTSR DRKDHMVRMH LYPADFRFDK PKKSRSPASA EAPGDSGERS EGEAMEICSE PVAASPAPAG ERRIYRHRIP STICFGQGAA RGFKSNKKKT KQC
May be involved in transcriptional regulation.
Specifications
Specifications
ZNF511
Monoclonal
Unconjugated
ZNF511
Liquid
ZNF511
ZNF511 (AAH19897.1, 20 a.a. ∽ 252 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
3G1
PBS with no preservative; pH 7.4
Q8NB15
ZNF511
Mouse
IgG2a, kappa
100 μg
-20° C
Primary
118472