Learn More
Aviva Systems Biology Corp ZNF526 Antibody - C-terminal region (ARP51699_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51699_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF526. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF526. Alternate names: DKFZp762O059, KIAA1951, MGC4267ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60591