Learn More
Abnova™ ZNF549 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00256051T01
Catalog No. 89-968-390
Add to cart
Description
Description
Specifications
Specifications
Western Blot
ZNF549 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 74.4kDa
Transient overexpression cell lysate was tested with anti-ZNF549 antibody (Mfr. No. H00256051-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
BC060863
pCMV-ZNF549 full-length
100 μL
Human