Learn More
Invitrogen™ ZNF549 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110605
Description
Immunogen sequence: MAEAALVNTP QIPMVTEEFV KPSQGHVTFE DIAVYFSQEE WGLLDEAQRC LYHDVMLENF SLMASVGCLH GIEAEEAPSE QTLSAQGVSQ ARTPKLGPSI PNAHSCEMCI LVMKDILYLS EHQGTLPWQK PYTSVASGKW FSFGSNLQQH QNQDSGEKHI RKEESSALLL NSCKIPLSDN LFPCKDVEKD FPTILGLLQH QTTHSRQEYA HRSRETFQQR RYKCEQVFNE KVHVTEHQRV HTGEKAYKRR EYGKSLNSKY LFVEHQRTHN AEKPYVCNIC.
ZNF549 may be involved in transcriptional regulation.
Specifications
ZNF549 | |
Polyclonal | |
Unconjugated | |
ZNF549 | |
Zinc finger protein 549; ZNF549 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
256051 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.09 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P9A3 | |
ZNF549 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ZNF549 (NP_001186224.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.