Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF550 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF550 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180424
|
Novus Biologicals
NBP180424 |
100 μL |
Each of 1 for $436.00
|
|
Description
ZNF550 Polyclonal specifically detects ZNF550 in Human samples. It is validated for Western Blot.Specifications
ZNF550 | |
Polyclonal | |
Purified | |
RUO | |
NP_001034743 | |
162972 | |
Synthetic peptide directed towards the middle region of human ZNF550. Peptide sequence CTQCGKAFHRSTYLIQHSVIHTGEMPYKCIECGKAFKRRSHLLQHQRVHT. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
MGC41917, zinc finger protein 550 | |
ZNF550 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title