Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF555 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF555 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180166
|
Novus Biologicals
NBP180166 |
100 μL |
Each of 1 for $436.00
|
|
Description
ZNF555 Polyclonal specifically detects ZNF555 in Human samples. It is validated for Western Blot.Specifications
ZNF555 | |
Polyclonal | |
Rabbit | |
NP_690004 | |
148254 | |
Synthetic peptide directed towards the N terminal of human ZNF555. Peptide sequence: QRDLYRDVMLETFQNLASVDDETQFKASGSVSQQDIYGEKIPKESKIATF | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
FLJ41894, MGC26707, zinc finger protein 555 | |
ZNF555 | |
IgG | |
Affinity Purified | |
73 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title