Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF566 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309367100UL
Description
ZNF566 Polyclonal specifically detects ZNF566 in Rat samples. It is validated for Western Blot.Specifications
ZNF566 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
FLJ14779, MGC12515, zinc finger protein 566 | |
The immunogen is a synthetic peptide directed towards the following sequence YECKECGKAFSSGSNFTQHQRIHTGEKPYECKECGNAFSQSSQLIKHQRI (NP_001128198). Peptide sequence YECKECGKAFSSGSNFTQHQRIHTGEKPYECKECGNAFSQSSQLIKHQRI | |
100 μg | |
Primary | |
Rat | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
84924 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction