Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF572 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18040620UL

 View more versions of this product

Catalog No. NBP18040620


Add to cart

Description

Description

ZNF572 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZNF572
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
ZNF572
Synthetic peptide directed towards the N terminal of human ZNF572. Peptide sequence HHNNSKADKLKEKPSEWSKRHRPQHYKHEDAKEMPLTWVQDEIWCHDSYE.
Immunogen affinity purified
RUO
Primary
137209
Western Blot
Western Blot 1:1000
NP_689625
FLJ38002, zinc finger protein 572
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit