Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
ZNF596 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS19528R
Catalog No. 50-198-4334
Description
Description
May be involved in transcriptional regulation.
Specifications
Specifications
ZNF596
Western Blot
Unconjugated
ZNF596
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
169270
ZNF596
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q8TC21
Zinc finger protein 596; ZNF596
KLH conjugated synthetic peptide derived from human ZNF596.
Protein A
RUO
Antibody
Polyclonal
Human