Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF597 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18040820UL

 View more versions of this product

Catalog No. NBP18040820


Add to cart

Description

Description

ZNF597 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZNF597
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
ZNF597
Synthetic peptide directed towards the N terminal of human ZNF597. Peptide sequence EDAALMGEEGKPEINQQLSLESMELDELALEKYPIAAPLVPYPEKSSEDG.
Immunogen affinity purified
RUO
Primary
146434
Western Blot
Western Blot 1:1000
NP_689670
FLJ33071, zinc finger protein 597
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit