Learn More
ZNF597 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 209221AP
Catalog No. 50-173-3158
Add to cart
Description
Description
May be involved in transcriptional regulation.
Specifications
Specifications
ZNF597
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96LX8
zinc finger protein 597, ZNF597
Rabbit
IgG
150 μL
-20°C
Primary
146434
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ZNF597
Liquid
ZNF597
ZNF597 Fusion Protein Ag15183
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human