Learn More
ZNF600 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 201001AP
Catalog No. 50-173-2697
Add to cart
Description
Description
This antibody is specific to ZNF600.
Consisting of around 63million baseswith over 1,400 genes,chromosome 19 makes up over 2% of human genomic DNA. Chromosome 19 includes a diversity of interesting genes and isrecognized for having the greatest gene density of the human chromosomes. It isthe genetic home for a number of immunoglobulin superfamily members including the killercell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAMand PSG family, and Fcalpha receptors. Key genesfor eye color and haircolor also map to chromosome 19. Peutz-Jegherssyndrome,spinocerebellar ataxia type 6, the stroke disorder CADASIL, hypercholesterolemia and Insulin-dependent diabetes have been linked to chromosome 19. Translocations with chromosome 19 and chromosome 14 can be seen in some lymphoproliferative disorders and typicallyinvolve the proto-oncogene Bcl-3.
Specifications
Specifications
ZNF600
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6ZNG1
KR ZNF1, zinc finger protein 600, ZNF600
Rabbit
IgG
150 μL
-20°C
Primary
162966, 667666
Western Blot
Unconjugated
ZNF600
Liquid
Zfp600, ZNF600
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse