Learn More
Aviva Systems Biology Corp ZNF618 Antibody - N-terminal region (ARP50760_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50760_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF618. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF618. Alternate names: RP11-56P10.2, FP13169, NEDD10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60426