Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF622 Mouse anti-Human, Clone: PCRP-ZNF622-1C11, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30748920UG
Description
ZNF622 Monoclonal specifically detects ZNF622 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
ZNF622 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552 | |
Recombinant full-length human ZNF622 protein | |
20 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Peptide Array | |
PCRP-ZNF622-1C11 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
90441 | |
Store at 4°C. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction