Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF622 Mouse anti-Human, Clone: PCRP-ZNF622-1C11, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen ZNF622
Clone PCRP-ZNF622-1C11
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122452
View Documents
Novus Biologicals
NBP30748920UG
20 μg
Each for $138.37
Add to cart
 
NB122451
View Documents
Novus Biologicals
NBP30748910UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

ZNF622 Monoclonal antibody specifically detects ZNF622 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

ZNF622
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
90441
IgG2a
Primary
Store at 4°C.
PCRP-ZNF622-1C11
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Human
MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552
Recombinant full-length human ZNF622 protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit