Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF645 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180410
Description
ZNF645 Polyclonal specifically detects ZNF645 in Human samples. It is validated for Western Blot.Specifications
ZNF645 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
NP_689790 | |
ZNF645 | |
Synthetic peptide directed towards the middle region of human ZNF645. Peptide sequence LSPQFTQTDAMDHRRWPAWKRLSPCPPTRSPPPSTLHGRSHHSHQRRHRR. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 92%. | |
Human, Canine, Yeast | |
IgG |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with 0.09% Sodium Azide | |
E3 ubiquitin ligase, HAKAIL, zinc finger protein 645 | |
Rabbit | |
Affinity Purified | |
RUO | |
158506 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title