Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF652 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537928
Description
This antibody is predicted to react with cow and pig based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
ZNF652 is a new candidate transcription factor.
Specifications
ZNF652 | |
Polyclonal | |
Unconjugated | |
ZNF652 | |
9530033F24Rik; DKFZp781E2122; KIAA0924; RGD1566329; Zfp652; Zinc finger protein 652; Znf652 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
22834 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9Y2D9 | |
ZNF652 | |
Peptide with sequence C-RHLAEKNSSAQHH, from the C-terminus of ZNF652 (aa594-606). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction