Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF662 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549168

Catalog No. PA549168

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF662
Polyclonal
PBS with 0.09% sodium azide
Q6ZS27
Zinc finger protein 662, ZNF662
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
389114
Western Blot
Unconjugated
ZNF662
Liquid
ZNF662
KLH conjugated synthetic peptide between 391-417 amino acids from the C-terminal region of human ZNF662.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit