Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF670 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF670 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180132
|
Novus Biologicals
NBP180132 |
100 μL |
Each of 1 for $436.00
|
|
Description
ZNF670 Polyclonal specifically detects ZNF670 in Human samples. It is validated for Western Blot.Specifications
ZNF670 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ12606, MGC12466, zinc finger protein 670 | |
ZNF670 | |
IgG | |
Affinity Purified | |
44 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_149990 | |
93474 | |
Synthetic peptide directed towards the N terminal of human ZNF670. Peptide sequence EIFRNLASVGNKSEDQNIQDDFKNPGRNLSSHVVERLFEIKEGSQYGETF. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title