Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF671 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304906100UL
Description
ZNF671 Polyclonal specifically detects ZNF671 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZNF671 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
79891 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry | |
Unconjugated | |
FLJ23506, zinc finger protein 671 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 265-534 of human ZNF671 (NP_079109.2). | |
100 μL | |
Primary | |
Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction