Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF671 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ZNF671
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095176
View Documents
Novus Biologicals
NBP304906100UL
100 μg
Each for $320.71
Add to cart
 
NB095177
View Documents
Novus Biologicals
NBP30490620UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

ZNF671 Polyclonal antibody specifically detects ZNF671 in Mouse, Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

ZNF671
Unconjugated
Rabbit
Mouse, Rat
FLJ23506, zinc finger protein 671
Recombinant fusion protein containing a sequence corresponding to amino acids 265-534 of human ZNF671 (NP_079109.2).
Polyclonal
Affinity purified
Immunohistochemistry
Purified
RUO
PBS with 50% glycerol, pH7.3.
79891
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit