Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF69 Mouse anti-Human, Biotin, Clone: OTI3C3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274954B

Catalog No. NBP274954B


Add to cart

Description

Description

ZNF69 Monoclonal antibody specifically detects ZNF69 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ZNF69
OTI3C3
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
ZNF69
Full length recombinant protein of human ZNF69 (NP_068734) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Biotin
PBS with 0.05% Sodium Azide
Cos5, hZNF3, MGC59928, zinc finger protein 69, zinc finger protein 69 (Cos5)
Mouse
IgG2b
0.1 mL
Primary
7620.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit