Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF690, Mouse anti-Human, Janelia Fluor 549, Clone: ZSCAN29/2610, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279881JF549
Description
ZNF690 Monoclonal antibody specifically detects ZNF690 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
ZNF690 | |
ZSCAN29/2610 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
ZSCAN29 | |
Recombinant full-length human ZNF690 protein | |
Protein A or G purified | |
RUO | |
Primary | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 2.1.1.43, EC 2.7.10.1, FLJ35867, MGC129894, MGC129895, Zfp690, zinc finger and SCAN domain containing 29, zinc finger and SCAN domain-containing protein 29, Zinc finger protein 690KOX31-like zinc finger protein, ZNF690 | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title