Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575924
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
May be involved in transcriptional regulation.
Specifications
ZNF7 | |
Polyclonal | |
Unconjugated | |
ZNF7 | |
C2-H2 type zinc finger protein; HF.16; KOX4; zf30; zinc finger protein 7; zinc finger protein 7 (KOX 4, clone HF.16); zinc finger protein HF.16; Zinc finger protein KOX4; zinc finger protein zfp30; Zinc finger protein-7 (KOX4); ZNF7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100910444, 7553 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P17097 | |
ZNF7 | |
Synthetic peptide corresponding to amino acids 32-77 of Human ZNF7. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction