Learn More
Aviva Systems Biology Corp ZNF708 Antibody - N-terminal region (ARP50662_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50662_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF708. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF708. Alternate names: DKFZp686L10267, FLJ16755, FLJ46310, KOX8, ZNF15, ZNF15L1ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-60404