Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF709 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF709 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180411
|
Novus Biologicals
NBP180411 |
100 μL |
Each of 1 for $436.00
|
|
Description
ZNF709 Polyclonal specifically detects ZNF709 in Human samples. It is validated for Western Blot.Specifications
ZNF709 | |
Polyclonal | |
Purified | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ38281, zinc finger protein 709 | |
ZNF709 | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
NP_689814 | |
163051 | |
Synthetic peptide directed towards the N terminal of human ZNF709. Peptide sequence DVMQETFVNLASIGENWEEKNIEDHKNQGRKLRSHMVERLCERKEGSQFG. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title