Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF710 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179434

 View more versions of this product

Catalog No. NBP179434


Add to cart

Description

Description

ZNF710 Polyclonal antibody specifically detects ZNF710 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot.
Specifications

Specifications

ZNF710
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
ZNF710
Synthetic peptide directed towards the N terminal of human ZNF710The immunogen for this antibody is ZNF710. Peptide sequence KFEKVEEEEQEVYEVSVPGDDKDAGPAEAPAEAASGGCDALVQSSAVKMI.
74 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Guinea pig 86%.
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat
Western Blot
Western Blot 1:1000
NP_940928
DKFZp547K1113, FLJ00306, FLJ37393, MGC163413, zinc finger protein 710
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
374655
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit