Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF710 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF710 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179434
|
Novus Biologicals
NBP179434 |
100 μL |
Each of 1 for $436.00
|
|
Description
ZNF710 Polyclonal specifically detects ZNF710 in Human samples. It is validated for Western Blot.Specifications
ZNF710 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
DKFZp547K1113, FLJ00306, FLJ37393, MGC163413, zinc finger protein 710 | |
ZNF710 | |
IgG | |
Affinity Purified | |
74 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_940928 | |
374655 | |
Synthetic peptide directed towards the N terminal of human ZNF710The immunogen for this antibody is ZNF710. Peptide sequence KFEKVEEEEQEVYEVSVPGDDKDAGPAEAPAEAASGGCDALVQSSAVKMI. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title