Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF711 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556530
Description
Immunogen sequence: DVVTDDGITL DHGLAAEVVH GPDIITETDV VTEGVIVPEA VLEADVAIEE DLEEDDGDHI LTSELITETV RVPEQVFVAD LVTGPNGHLE HVVQDCVSGV DSPTMVSEEV LVTN Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
This gene encodes a zinc finger protein of unknown function. It bears similarity to a zinc finger protein which acts as a transcriptional activator. This gene lies in a region of the X chromosome which has been associated with mental retardation.
Specifications
ZNF711 | |
Polyclonal | |
Unconjugated | |
ZNF711 | |
2810409C01Rik; A230078I01Rik; CMPX1; dJ75N13.1; dJ75N13.1 (znf6-like); MRX97; Zfp711; Zinc finger protein 6; zinc finger protein 6 (CMPX1); zinc finger protein 711; ZNF4; ZNF5; ZNF6; ZNF711 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7552 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y462 | |
ZNF711 | |
Recombinant protein corresponding to Human ZNF711. Recombinant protein control fragment (Product #RP-93691). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction