Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF747 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549051

Catalog No. PA549051

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ZNF747
Polyclonal
PBS with 0.09% sodium azide
Q9BV97
KRAB domain-containing protein ZNF747, ZNF747
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65988
Western Blot
Unconjugated
ZNF747
Liquid
ZNF747
KLH conjugated synthetic peptide between 156-182 amino acids from the C-terminal region of human ZNF747.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit