Learn More
ZNF747 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 199011AP
Catalog No. 50-173-2591
Add to cart
Description
Description
ZNF747 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications
Specifications
ZNF747
0.18 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BV97
zinc finger protein 747, ZNF747
Rabbit
IgG
150 μL
-20°C
Primary
65988
Western Blot
Unconjugated
ZNF747
Liquid
ZNF747
ZNF747 Fusion Protein Ag13821
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human