Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF75A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549009

Catalog No. PA549009

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF75A
Polyclonal
PBS with 0.09% sodium azide
Q96N20
Zinc finger protein 75A, ZNF75A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7627
Western Blot
Unconjugated
ZNF75A
Liquid
ZNF75A
KLH conjugated synthetic peptide between 98-124 amino acids from the Central region of human ZNF75A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit