Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF75A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF75A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180172
|
Novus Biologicals
NBP180172 |
100 μL |
Each of 1 for $436.00
|
|
Description
ZNF75A Polyclonal specifically detects ZNF75A in Human samples. It is validated for Western Blot.Specifications
ZNF75A | |
Polyclonal | |
Rabbit | |
Human | |
FLJ31529, MGC59959, zinc finger protein 75a | |
ZNF75A | |
IgG | |
Affinity Purified | |
35 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_694573 | |
7627 | |
Synthetic peptide directed towards the N terminal of human ZNF75A. Peptide sequence FVLPKPKVISCLEQGEEPWVQVSPEFKDSAGKSPTGLKLKNDTENHQPVS. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title