Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF768 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549038

Catalog No. PA549038

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and mouse based on sequence homology.

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF768
Polyclonal
PBS with 0.09% sodium azide
Q9H5H4
Zinc finger protein 768, ZNF768
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79724
Western Blot
Unconjugated
ZNF768
Liquid
ZNF768
KLH conjugated synthetic peptide between 3-29 amino acids from the N-terminal region of human ZNF768.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit