Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF775 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549197

Catalog No. PA549197


Add to cart

Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF775
Polyclonal
Unconjugated
ZNF775
Liquid
ZNF775
KLH conjugated synthetic peptide between 406-432 amino acids from the C-terminal region of human ZNF775.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q96BV0
Zinc finger protein 775, ZNF775
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
285971
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit