Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF775 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | ZNF775 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB123337
|
Novus Biologicals
NBP309218100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
ZNF775 Polyclonal specifically detects ZNF775 in Human samples. It is validated for Western Blot.Specifications
ZNF775 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
ZNF775 zinc finger protein 775 | |
The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF775 (NP_775951). Peptide sequence MESGLAGNGTGAGLVMKVKQEKPERLLQTLAPQAMLVEKDKENIFQQHRG | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
285971 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title