Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF780B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF780B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179348
|
Novus Biologicals
NBP179348 |
100 μL |
Each of 1 for $436.00
|
|
Description
ZNF780B Polyclonal specifically detects ZNF780B in Human samples. It is validated for Western Blot.Specifications
ZNF780B | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
zinc finger protein 780B | |
ZNF780B | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_001005851 | |
163131 | |
Synthetic peptide directed towards the N terminal of human ZNF780BThe immunogen for this antibody is ZNF780B. Peptide sequence RWYPDLESKYGPEKISPENDIFEINLPKHVIKQISKTLGLEAFYFRNDSE. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title