Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF79 Mouse anti-Human, DyLight™ 755, Clone: OTI3D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274955IR

Catalog No. NBP274955IR


Add to cart

Description

Description

ZNF79 Monoclonal antibody specifically detects ZNF79 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF79
OTI3D2
Western Blot
Affinity Purified, Conjugated
ZNF79
Full length recombinant protein of human ZNF79 (NP_009066) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
pT7, zinc finger protein 79, zinc finger protein 79 (pT7), ZNFpT7
Mouse
IgG2b
0.1 mL
Primary
7633.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit