Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF791 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569054
Description
This target displays homology in the following species: Horse: 77%; Human: 100%; Mouse: 82%; Pig: 75%; Rat: 75%.
ZNF791 may be involved in transcriptional regulation.
Specifications
ZNF791 | |
Polyclonal | |
Unconjugated | |
ZNF791 | |
Zinc finger protein 791; ZNF791 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
163049 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3KP31 | |
ZNF791 | |
synthetic peptide directed towards the N terminal region of human ZNF791. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction