Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF81 Mouse anti-Human, Alexa Fluor 532, Clone: PCRP-ZNF81-2G2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314088AF532
This item is not returnable.
View return policy
Description
ZNF81 Monoclonal antibody specifically detects ZNF81 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ZNF81 | |
Monoclonal | |
Alexa Fluor 532 | |
dJ54B20.6, FLJ44367, HFZ20X-linked 45, zinc finger protein 81, zinc finger protein 81 (HFZ20) | |
Recombinant full-length human ZNF81 protein (Uniprot: P51508) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-ZNF81-2G2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
347344 | |
Store at 4°C in the dark. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title