Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF821 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | ZNF821 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180335
|
Novus Biologicals
NBP180335 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
ZNF821 Polyclonal specifically detects ZNF821 in Human samples. It is validated for Western Blot.Specifications
ZNF821 | |
Polyclonal | |
Rabbit | |
NP_060000 | |
55565 | |
Synthetic peptide directed towards the C terminal of human LOC55565. Peptide sequence LRAQFGQDPSAMAALAAEMNFFQLPVSGVELDSQLLGKMAFEEQNSSSLH. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
FLJ16350, zinc finger protein 821 | |
ZNF821 | |
IgG |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title