Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF827 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $378.35

Specifications

Antigen ZNF827
Immunogen Synthetic peptide directed towards the middle region of human LOC152485. Peptide sequence VCKTKNMFERHLQIHLITRMFECDVCHKFMKTPEQLLEHKKCHTVPTGGL.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1804320 View Documents Novus Biologicals
NBP18043120UL
20ul Each for $152.22
Add to cart
 
NBP180431 View Documents Novus Biologicals
NBP180431
100 ul Each for $378.35
Add to cart
 
Description

Description

ZNF827 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZNF827
Rabbit
Polyclonal
Unconjugated
152485
Affinity Purified
Western Blot
RUO
Synthetic peptide directed towards the middle region of human LOC152485. Peptide sequence VCKTKNMFERHLQIHLITRMFECDVCHKFMKTPEQLLEHKKCHTVPTGGL.
Primary
IgG
zinc finger protein 827
PBS & 2% Sucrose. with No Preservative
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit