Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF879 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564375
Description
Immunogen sequence: LEQGEDPWMV ESGVPQGAHL GWESLFGTIV SKEENQEVMK KLIIDGTFDF KLEKTYINED KLEKQQGKKN RLFSKVLVTI K Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 49%.
The exact functions of DKFZp686E2433 remain unknown.
Specifications
ZNF879 | |
Polyclonal | |
Unconjugated | |
ZNF879 | |
DKFZp686E2433; Zinc finger protein 879; ZNF879 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
345462 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
B4DU55 | |
ZNF879 | |
Recombinant protein corresponding to Human ZNF879. Recombinant protein control fragment (Product #RP-104175). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction