Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF879 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543719
Description
Peptide sequence: ATGNVSELGV PLASCAVWSC APPAEAGPGA FSKRGSREGE MARRLLPAHV.
The exact functions of DKFZp686E2433 remain unknown.
Specifications
ZNF879 | |
Polyclonal | |
Unconjugated | |
ZNF879 | |
DKFZp686E2433; Zinc finger protein 879; ZNF879 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
345462 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
B4DU55 | |
ZNF879 | |
Synthetic peptide directed towards the N-terminal of human DKFZp686E2433. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction