Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF9 Polyclonal Antibody
ZNF9 Polyclonal Antibody
Supplier: Thermo Scientific PA5104287
Description
ZNF9 Polyclonal Antibody for Western Blot

Specifications
ZNF9 | |
Polyclonal | |
Unconjugated | |
CNBP | |
AA408710; CCHC-type zinc finger nucleic acid binding protein; CCHC-type zinc finger, nucleic acid binding protein; cellular nucleic acid binding protein; cellular nucleic acid binding protein 1; cellular nucleic acid-binding protein; CNBP; CNBP1; DM2; erythroid differentiation-related; HGNC:13164; PROMM; RNF163; sterol regulatory element-binding protein; ZCCHC22; zinc finger protein 273; zinc finger protein 9; zinc finger protein 9 (a cellular retroviral nucleic acid binding protein); Znf9 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12785, 64530, 7555 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P53996, P62633, P62634 | |
CNBP | |
A synthesized peptide derived from human CNBP(Accession P62633), corresponding to amino acid residues R11-G61. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction