Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF92 Monoclonal Antibody (1F2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524530
Description
Peptide Sequence: FNQSSIFTKH KIIHTEGKSY KCEKCGNAFN QSSNLTARKI IYTGEKPYKY EECDKAFNKF STLITHQIIY TGEKPCKHEC GRAFNKSSNY TKEKLQT
ZNF92 Monoclonal antibody specifically detects ZNF92 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
ZNF92 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q03936 | |
ZNF92 | |
ZNF92 (NP_689839, 490 a.a. ∽ 586 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1F2 | |
Unconjugated | |
ZNF92 | |
epididymis luminal protein 203; HEL-203; HPF12; HTF12; TF12; zinc finger protein 92; Zinc finger protein HTF12; ZNF92 | |
Mouse | |
Protein A | |
RUO | |
168374 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction