Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF96 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen ZNF96
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125970
View Documents
Novus Biologicals
NBP31053425UL
25 μg
Each for $106.19
Add to cart
 
NB125969
View Documents
Novus Biologicals
NBP310534100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

ZNF96 Polyclonal antibody specifically detects ZNF96 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

ZNF96
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
9753
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Mouse
KIAA0426dJ29K1.2, ZFP96, zinc finger and SCAN domain containing 12, Zinc finger protein 305ZNF305, Zinc finger protein 96ZNF96zinc finger and SCAN domain-containing protein 12, ZNF29K1
The immunogen is a synthetic peptide corresponding to a region of Mouse (NP_057893). Peptide sequence WEGTEAEQNPASRLAKDALECEEAHNPGEESSGISHEDSQPLRNENGVNS
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit