Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNHIT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529483
Description
Recommended positive controls: mouse testis. Predicted reactivity: Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
ZNHIT2 | |
Polyclonal | |
Unconjugated | |
Znhit2 | |
C11orf5; FON; ORF6; Protein FON; RGD1307918; zinc finger HIT domain-containing protein 2; zinc finger HIT-type containing 2; zinc finger, HIT domain containing 2; zinc finger, HIT domain containing 2, pseudogene; zinc finger, HIT type 2; zinc finger, HIT-type containing 2; Znhit2; Znhit2-ps | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29805, 741 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9QY66, Q9UHR6 | |
Znhit2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 273 of Human ZNHIT2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction